Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BMP6
Entrez Gene ID
654
Gene Name
bone morphogenetic protein 6
Gene Aliases
VGR, VGR1
Chromosome Location
Chr.6: 7726778 - 7881728 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001718.4 NP_001709.1 6 - 7 1568 74
GenBank mRNA AK300628.1 - 7 - 8 1528 74
BC096084.2 - 7 - 8 1136 74
BC096085.2 - 7 - 8 1136 74
BC099624.1 - 7 - 8 1136 74
BC099625.1 - 7 - 8 1300 74
BC111763.1 - 7 - 8 1300 74
BC112352.1 - 7 - 8 1300 74
BC113018.1 - 7 - 8 1195 74
BC113019.1 - 7 - 8 1192 74
BC128031.1 - 7 - 8 1092 74
M60315.1 - 6 - 7 1549 74

View Full Product Details