Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RALB
Entrez Gene ID
5899
Gene Name
RALB Ras like proto-oncogene B
Gene Aliases
-
Chromosome Location
Chr.2: 120240070 - 120294711 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002881.2 NP_002872.1 1 - 2 145 100
XM_011511574.1 XP_011509876.1 1 - 2 133 100
GenBank mRNA AK127675.1 - 1 - 2 130 100
BQ893549.1 - - 150 100
M35416.1 - 1 - 2 125 100

View Full Product Details