The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002690.2 | NP_002681.1 | 13 - 14 | 1078 | 129 |
XM_005273535.3 | XP_005273592.1 | 14 - 15 | 1180 | 129 | |
XM_005273536.3 | XP_005273593.1 | 13 - 14 | 1093 | 129 | |
XM_005273537.3 | XP_005273594.1 | 12 - 13 | 988 | 129 | |
XM_005273538.2 | XP_005273595.1 | 13 - 14 | 1090 | 129 | |
XM_005273539.2 | XP_005273596.1 | 12 - 13 | 985 | 129 | |
XM_005273540.4 | XP_005273597.1 | 9 - 10 | 839 | 129 | |
XM_006716353.2 | XP_006716416.1 | 10 - 11 | 724 | 129 | |
XM_017013583.1 | XP_016869072.1 | 11 - 12 | 982 | 129 | |
XM_017013584.1 | XP_016869073.1 | 10 - 11 | 875 | 129 | |
GenBank mRNA | AK223537.1 | - | 13 - 14 | 1077 | 129 |
AK294025.1 | - | 9 - 10 | 797 | 129 | |
AK314976.1 | - | 13 - 14 | 1039 | 129 | |
BC100288.1 | - | 13 - 14 | 1042 | 129 | |
BC106909.1 | - | 13 - 14 | 1004 | 129 | |
BM678092.1 | - | 1 - 2 | 129 | ||
CR536503.1 | - | 13 - 14 | 908 | 129 | |
CR541802.1 | - | 13 - 14 | 908 | 129 | |
D29013.1 | - | 13 - 14 | 1021 | 129 | |
L11607.1 | - | 13 - 14 | 908 | 129 |