Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELN
Entrez Gene ID
2006
Gene Name
elastin
Gene Aliases
SVAS, WBS, WS
Chromosome Location
Chr.7: 74027772 - 74069907 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000501.3 NP_000492.2 5 - 6 633 151
NM_001081752.2 NP_001075221.1 4 - 5 603 151
NM_001081753.2 NP_001075222.1 5 - 6 633 151
NM_001081754.2 NP_001075223.1 5 - 6 633 151
NM_001081755.2 NP_001075224.1 5 - 6 633 151
NM_001278912.1 NP_001265841.1 5 - 6 633 151
NM_001278914.1 NP_001265843.1 4 - 5 603 151
NM_001278915.1 NP_001265844.1 5 - 6 633 151
NM_001278916.1 NP_001265845.1 5 - 6 633 151
NM_001278917.1 NP_001265846.1 4 - 5 603 151
NM_001278918.1 NP_001265847.1 4 - 5 603 151
NM_001278939.1 NP_001265868.1 5 - 6 633 151
XM_005250188.1 XP_005250245.1 5 - 6 308 151
XM_011515868.1 XP_011514170.1 5 - 6 319 151
XM_011515869.1 XP_011514171.1 4 - 5 248 151
XM_011515871.1 XP_011514173.1 5 - 6 319 151
XM_011515872.1 XP_011514174.1 5 - 6 319 151
XM_011515873.1 XP_011514175.1 5 - 6 319 151
XM_011515876.1 XP_011514178.1 5 - 6 319 151
XM_011515877.1 XP_011514179.1 5 - 6 319 151
XM_017011813.1 XP_016867302.1 4 - 5 219 151
XM_017011814.1 XP_016867303.1 5 - 6 319 151
GenBank mRNA AB208942.1 - 4 - 5 229 151
AK092232.1 - 4 - 5 218 151
AK095990.1 - 5 - 6 253 151
AK125511.1 - 4 - 5 263 151
AK225659.1 - 5 - 6 247 151
AK304845.1 - 5 - 6 633 151
BC035570.1 - 5 - 6 238 151
BC050379.1 - 5 - 6 251 151
BC065566.1 - 5 - 6 264 151
BP284909.1 - 5 - 6 325 151
BX537939.1 - 5 - 6 253 151
BX538199.1 - 4 - 5 235 151
M36860.1 - 5 - 6 283 151
X52896.1 - 5 - 6 282 151

View Full Product Details