The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040260.3 | NP_001035350.2 | 1 - 2 | 1170 | 69 |
NM_001040261.4 | NP_001035351.4 | 1 - 2 | 1170 | 69 | |
NR_036614.1 | 1 - 2 | 1170 | 69 | ||
XM_005262785.2 | XP_005262842.1 | 1 - 2 | 999 | 69 | |
XM_005262786.2 | XP_005262843.1 | 1 - 2 | 999 | 69 | |
XM_017007832.1 | XP_016863321.1 | 1 - 2 | 999 | 69 | |
XM_017007833.1 | XP_016863322.1 | 1 - 2 | 999 | 69 | |
GenBank mRNA | AB209181.1 | - | 1 - 2 | 1170 | 69 |
AL834498.1 | - | 1 - 2 | 217 | 69 | |
BC032726.1 | - | 1 - 2 | 463 | 69 |