The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006342.2 | NP_006333.1 | 3 - 4 | 463 | 63 |
XM_005247929.2 | XP_005247986.1 | 3 - 4 | 461 | 63 | |
XM_011513386.2 | XP_011511688.1 | 3 - 4 | 461 | 63 | |
XM_017007653.1 | XP_016863142.1 | 3 - 4 | 515 | 63 | |
GenBank mRNA | AF093543.1 | - | 3 - 4 | 416 | 63 |
AJ243997.1 | - | 3 - 4 | 408 | 63 | |
AK225631.1 | - | 3 - 4 | 431 | 63 | |
AK302463.1 | - | 3 - 4 | 434 | 63 | |
AK304579.1 | - | 2 - 3 | 192 | 63 | |
AK310660.1 | - | 2 - 3 | 186 | 63 | |
BC106071.1 | - | 3 - 4 | 424 | 63 | |
BC111771.1 | - | 3 - 4 | 376 | 63 | |
DB098941.1 | - | 3 - 4 | 461 | 63 |