The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016141.3 | NP_057225.2 | 7 - 8 | 1078 | 138 |
GenBank mRNA | AF078849.1 | - | 7 - 8 | 1054 | 138 |
AK001081.1 | - | 4 - 5 | 574 | 138 | |
AK074348.1 | - | 7 - 8 | 1062 | 138 | |
AK222653.1 | - | 7 - 8 | 1014 | 138 | |
AK293253.1 | - | 5 - 6 | 714 | 138 | |
BC131620.1 | - | 7 - 8 | 1024 | 138 | |
BX649189.1 | - | 7 - 8 | 897 | 138 |