The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002668.2 | NP_002659.1 | 1 - 2 | 265 | 68 |
GenBank mRNA | BC109066.2 | - | 1 - 2 | 166 | 68 |
DB220888.1 | - | 1 - 2 | 266 | 68 | |
L09604.1 | - | 1 - 2 | 176 | 68 |