The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002668.2 | NP_002659.1 | 2 - 3 | 411 | 84 |
GenBank mRNA | BC109066.2 | - | 2 - 3 | 312 | 84 |
L09604.1 | - | 2 - 3 | 322 | 84 |