The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282167.1 | NP_001269096.1 | 3 - 4 | 660 | 64 |
NM_022825.3 | NP_073736.2 | 3 - 4 | 515 | 64 | |
NM_203473.2 | NP_982299.1 | 3 - 4 | 515 | 64 | |
NM_203474.1 | NP_982300.1 | 2 - 3 | 370 | 64 | |
NM_203475.2 | NP_982301.1 | 3 - 4 | 515 | 64 | |
XM_005272635.2 | XP_005272692.1 | 3 - 4 | 870 | 64 | |
XM_005272636.2 | XP_005272693.1 | 3 - 4 | 870 | 64 | |
XM_005272637.1 | XP_005272694.1 | 4 - 5 | 797 | 64 | |
XM_006724545.3 | XP_006724608.1 | 4 - 5 | 580 | 64 | |
XM_006724546.3 | XP_006724609.1 | 4 - 5 | 652 | 64 | |
XM_006724547.1 | XP_006724610.1 | 3 - 4 | 372 | 64 | |
XM_011543948.2 | XP_011542250.1 | 3 - 4 | 799 | 64 | |
XM_017029735.1 | XP_016885224.1 | 3 - 4 | 880 | 64 | |
XM_017029736.1 | XP_016885225.1 | 3 - 4 | 372 | 64 | |
GenBank mRNA | AF317058.1 | - | 2 - 3 | 336 | 64 |
AF317059.1 | - | 2 - 3 | 336 | 64 | |
AF317060.1 | - | 2 - 3 | 336 | 64 | |
AF317061.1 | - | 2 - 3 | 336 | 64 | |
AK056340.1 | - | 3 - 4 | 829 | 64 | |
AK057727.1 | - | 3 - 4 | 372 | 64 | |
AK293347.1 | - | 3 - 4 | 625 | 64 | |
AK314745.1 | - | 2 - 3 | 385 | 64 | |
AK316378.1 | - | 2 - 3 | 407 | 64 | |
AL579208.3 | - | 3 - 4 | 434 | 64 | |
BC019080.2 | - | 3 - 4 | 462 | 64 | |
BM543485.1 | - | 3 - 4 | 486 | 64 | |
BM545893.1 | - | - | 386 | 64 | |
L08239.1 | - | 1 - 2 | 252 | 64 |