Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS6
Entrez Gene ID
9628
Gene Name
regulator of G-protein signaling 6
Gene Aliases
GAP
Chromosome Location
Chr.14: 71932100 - 72632478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204416.2 NP_001191345.1 13 - 14 1254 78
NM_001204417.2 NP_001191346.1 13 - 14 1254 78
NM_001204418.2 NP_001191347.1 13 - 14 1254 78
NM_001204423.1 NP_001191352.1 12 - 13 1019 78
NM_001204424.1 NP_001191353.1 13 - 14 1492 78
NM_004296.6 NP_004287.3 13 - 14 1254 78
NR_135235.1 13 - 14 1254 78
XM_011537393.2 XP_011535695.1 13 - 14 1639 78
XM_011537397.1 XP_011535699.1 11 - 12 809 78
XM_011537407.2 XP_011535709.1 14 - 15 1725 78
XM_017021818.1 XP_016877307.1 13 - 14 1154 78
XM_017021819.1 XP_016877308.1 13 - 14 1154 78
XM_017021821.1 XP_016877310.1 12 - 13 989 78
XM_017021822.1 XP_016877311.1 12 - 13 989 78
XM_017021823.1 XP_016877312.1 12 - 13 989 78
XM_017021825.1 XP_016877314.1 12 - 13 989 78
XM_017021826.1 XP_016877315.1 12 - 13 989 78
XM_017021827.1 XP_016877316.1 12 - 13 989 78
XM_017021828.1 XP_016877317.1 12 - 13 989 78
XM_017021829.1 XP_016877318.1 13 - 14 1154 78
XM_017021830.1 XP_016877319.1 12 - 13 989 78
XM_017021831.1 XP_016877320.1 12 - 13 989 78
XM_017021832.1 XP_016877321.1 12 - 13 989 78
XM_017021833.1 XP_016877322.1 12 - 13 989 78
XM_017021834.1 XP_016877323.1 11 - 12 809 78
GenBank mRNA AB209462.1 - 13 - 14 1060 78
AF073921.1 - 8 - 9 617 78
AF107619.1 - 12 - 13 969 78
AF107620.1 - 12 - 13 969 78
AF156932.1 - 12 - 13 969 78
AK294486.1 - 13 - 14 1492 78
AK294895.1 - 12 - 13 1019 78
AK302319.1 - 11 - 12 865 78
AK313481.1 - 13 - 14 1314 78
AY309097.1 - 12 - 13 969 78
AY309098.1 - 12 - 13 969 78
AY309099.1 - 12 - 13 969 78
AY309100.1 - 12 - 13 969 78
BC104877.1 - 13 - 14 1147 78
BC113572.1 - 13 - 14 1147 78
BC143699.1 - 13 - 14 1147 78

View Full Product Details