The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000400.3 | NP_000391.1 | 1 - 2 | 55 | 80 |
XM_011526611.2 | XP_011524913.1 | 1 - 2 | 115 | 80 | |
GenBank mRNA | BC110522.1 | - | 1 - 2 | 36 | 80 |
BC110523.1 | - | 1 - 2 | 36 | 80 | |
BX376468.2 | - | 1 - 2 | 39 | 80 |