The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001664.2 | NP_001001664.1 | 3 - 4 | 604 | 110 |
XM_005263655.4 | XP_005263712.1 | 3 - 4 | 470 | 110 | |
GenBank mRNA | BC071613.1 | - | 3 - 4 | 583 | 110 |