Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC8
Entrez Gene ID
123016
Gene Name
tetratricopeptide repeat domain 8
Gene Aliases
BBS8, RP51
Chromosome Location
Chr.14: 88824153 - 88877996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144596.3 NP_653197.2 7 - 8 774 149
NM_198309.3 NP_938051.1 6 - 7 744 149
XM_006720035.1 XP_006720098.1 5 - 6 1026 149
XM_017020977.1 XP_016876466.1 5 - 6 1026 149
GenBank mRNA AK124675.1 - 6 - 7 624 149
AK296077.1 - 4 - 5 833 149
AK315772.1 - 5 - 6 629 149
AY366523.1 - 5 - 6 600 149
AY373972.1 - 5 - 6 600 149
BC001563.1 - 1 - 2 92 149
BC095433.1 - 5 - 6 582 149
BX247959.1 - 7 - 8 898 149
KU178850.1 - 5 - 6 548 149

View Full Product Details