The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170634.1 | NP_001164105.1 | 7 - 8 | 904 | 60 |
NM_001170937.1 | NP_001164408.1 | 7 - 8 | 895 | 60 | |
NM_004960.3 | NP_004951.1 | 7 - 8 | 907 | 60 | |
XM_011545781.1 | XP_011544083.1 | 7 - 8 | 873 | 60 | |
GenBank mRNA | AK098774.1 | - | 7 - 8 | 875 | 60 |
AK290936.1 | - | 7 - 8 | 875 | 60 | |
AK299129.1 | - | 7 - 8 | 663 | 60 | |
BC000402.2 | - | 7 - 8 | 870 | 60 | |
BC002459.1 | - | 7 - 8 | 842 | 60 | |
BC026062.2 | - | 7 - 8 | 860 | 60 | |
BT007131.1 | - | 6 - 7 | 802 | 60 | |
CR456747.1 | - | 6 - 7 | 802 | 60 | |
S62140.1 | - | 7 - 8 | 880 | 60 | |
U36561.1 | - | 6 - 7 | 811 | 60 | |
X71428.1 | - | 7 - 8 | 871 | 60 |