Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAT8
Entrez Gene ID
84148
Gene Name
lysine acetyltransferase 8
Gene Aliases
MOF, MYST1, ZC2HC8, hMOF
Chromosome Location
Chr.16: 31117664 - 31131393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032188.2 NP_115564.2 2 - 3 302 77
NM_182958.2 NP_892003.2 2 - 3 302 77
XM_011545970.1 XP_011544272.1 3 - 4 843 77
XM_011545971.1 XP_011544273.1 3 - 4 848 77
GenBank mRNA AF217501.1 - 2 - 3 268 77
AF260665.2 - 2 - 3 200 77
AK021872.1 - 2 - 3 299 77
AK024102.1 - 2 - 3 300 77
AK225660.1 - 2 - 3 302 77
AK291106.1 - 2 - 3 299 77
BC037773.1 - 2 - 3 299 77
BP342476.1 - 2 - 3 302 77

View Full Product Details