The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022823.2 | NP_073734.1 | 4 - 5 | 860 | 59 |
XM_005264499.3 | XP_005264556.1 | 4 - 5 | 904 | 59 | |
GenBank mRNA | AK026015.1 | - | 4 - 5 | 820 | 59 |
AY358136.1 | - | 4 - 5 | 578 | 59 | |
BC032725.1 | - | 3 - 4 | 491 | 59 |