The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042692.2 | NP_001036157.1 | 3 - 4 | 471 | 92 |
NM_015910.5 | NP_056994.3 | 9 - 10 | 1286 | 92 | |
NR_122106.1 | 3 - 4 | 471 | 92 | ||
XM_005264348.3 | XP_005264405.1 | 9 - 10 | 1326 | 92 | |
XM_011532881.2 | XP_011531183.1 | 10 - 11 | 1110 | 92 | |
XM_011532884.2 | XP_011531186.1 | 9 - 10 | 1326 | 92 | |
XM_011532886.2 | XP_011531188.1 | 9 - 10 | 1326 | 92 | |
XM_011532887.2 | XP_011531189.1 | 9 - 10 | 1326 | 92 | |
XM_011532890.2 | XP_011531192.1 | 9 - 10 | 1326 | 92 | |
XM_011532891.2 | XP_011531193.1 | 10 - 11 | 1311 | 92 | |
XM_017004251.1 | XP_016859740.1 | 11 - 12 | 1964 | 92 | |
XM_017004252.1 | XP_016859741.1 | 10 - 11 | 1110 | 92 | |
XM_017004253.1 | XP_016859742.1 | 9 - 10 | 1326 | 92 | |
XM_017004254.1 | XP_016859743.1 | 9 - 10 | 1326 | 92 | |
GenBank mRNA | AF131737.1 | - | 3 - 4 | 471 | 92 |
AK001849.1 | - | 1 - 2 | 186 | 92 | |
AK022647.1 | - | 3 - 4 | 445 | 92 | |
AK055030.1 | - | 10 - 11 | 1311 | 92 | |
AK309028.1 | - | 4 - 5 | 526 | 92 | |
AK310290.1 | - | 9 - 10 | 953 | 92 | |
AL833623.1 | - | 9 - 10 | 1072 | 92 | |
BC093752.1 | - | 3 - 4 | 416 | 92 | |
BC093754.1 | - | 3 - 4 | 416 | 92 | |
BX538331.1 | - | 9 - 10 | 1032 | 92 | |
BX647132.1 | - | 3 - 4 | 3799 | 92 |