The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000932.2 | NP_000923.1 | 25 - 26 | 3165 | 100 |
NM_001184883.1 | NP_001171812.1 | 23 - 24 | 2964 | 100 | |
NM_001316314.1 | NP_001303243.1 | 25 - 26 | 3165 | 100 | |
XM_011545101.2 | XP_011543403.1 | 25 - 26 | 3153 | 100 | |
GenBank mRNA | BC032659.1 | - | 23 - 24 | 2911 | 100 |
BC041625.1 | - | 7 - 8 | 828 | 100 | |
BC142681.1 | - | 25 - 26 | 3118 | 100 | |
BC146645.1 | - | 25 - 26 | 3141 | 100 | |
Z16411.1 | - | 19 - 20 | 2490 | 100 | |
Z26649.1 | - | 25 - 26 | 3168 | 100 |