The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000932.2 | NP_000923.1 | 7 - 8 | 724 | 88 |
NM_001184883.1 | NP_001171812.1 | 5 - 6 | 523 | 88 | |
NM_001316314.1 | NP_001303243.1 | 7 - 8 | 724 | 88 | |
XM_011545101.2 | XP_011543403.1 | 7 - 8 | 712 | 88 | |
XM_017017925.1 | XP_016873414.1 | 7 - 8 | 712 | 88 | |
GenBank mRNA | BC032659.1 | - | 5 - 6 | 470 | 88 |
BC142681.1 | - | 7 - 8 | 677 | 88 | |
BC146645.1 | - | 7 - 8 | 700 | 88 | |
Z16411.1 | - | 1 - 2 | 49 | 88 | |
Z26649.1 | - | 7 - 8 | 727 | 88 |