The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144664.4 | NP_653265.3 | 3 - 4 | 519 | 86 |
XM_005273775.4 | XP_005273832.1 | 3 - 4 | 861 | 86 | |
XM_005273776.4 | XP_005273833.1 | 3 - 4 | 902 | 86 | |
XM_011542610.2 | XP_011540912.1 | 4 - 5 | 293 | 86 | |
XM_011542611.2 | XP_011540913.1 | 3 - 4 | 860 | 86 | |
XM_011542612.1 | XP_011540914.1 | 3 - 4 | 361 | 86 | |
XM_011542613.1 | XP_011540915.1 | 3 - 4 | 254 | 86 | |
GenBank mRNA | AK021591.1 | - | 2 - 3 | 1339 | 86 |
AK313727.1 | - | 3 - 4 | 260 | 86 | |
BC026013.2 | - | 3 - 4 | 519 | 86 | |
BC028727.1 | - | 3 - 4 | 351 | 86 | |
BX647586.1 | - | 3 - 4 | 228 | 86 |