The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012216.3 | NP_036348.2 | 4 - 5 | 1495 | 70 |
NM_052817.2 | NP_438112.2 | 4 - 5 | 1495 | 70 | |
XM_005262062.4 | XP_005262119.1 | 4 - 5 | 5758 | 70 | |
XM_017029239.1 | XP_016884728.1 | 4 - 5 | 5758 | 70 | |
GenBank mRNA | AF196481.1 | - | 4 - 5 | 935 | 70 |
AY625004.1 | - | 3 - 4 | 862 | 70 | |
BC017707.1 | - | 3 - 4 | 889 | 70 | |
BT006663.1 | - | 3 - 4 | 862 | 70 | |
Y18880.1 | - | 4 - 5 | 1119 | 70 |