The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012216.3 | NP_036348.2 | 9 - 10 | 2380 | 144 |
NM_052817.2 | NP_438112.2 | 9 - 10 | 2290 | 144 | |
XM_005262062.4 | XP_005262119.1 | 9 - 10 | 6643 | 144 | |
XM_017029239.1 | XP_016884728.1 | 9 - 10 | 6553 | 144 | |
GenBank mRNA | AF196481.1 | - | 9 - 10 | 1730 | 144 |
AY625004.1 | - | 8 - 9 | 1657 | 144 | |
BC017707.1 | - | 8 - 9 | 1684 | 144 | |
BT006663.1 | - | 8 - 9 | 1657 | 144 | |
Y18880.1 | - | 9 - 10 | 2004 | 144 |