The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145062.2 | NP_659499.2 | 8 - 9 | 1731 | 138 |
XM_011535565.2 | XP_011533867.1 | 8 - 9 | 2124 | 138 | |
XM_011535567.1 | XP_011533869.1 | 8 - 9 | 1408 | 138 | |
XM_017010393.1 | XP_016865882.1 | 8 - 9 | 1703 | 138 | |
XM_017010394.1 | XP_016865883.1 | 7 - 8 | 1157 | 138 | |
XM_017010395.1 | XP_016865884.1 | 9 - 10 | 2261 | 138 | |
XM_017010396.1 | XP_016865885.1 | 9 - 10 | 1641 | 138 | |
GenBank mRNA | AK054582.1 | - | 8 - 9 | 1717 | 138 |
BC016879.1 | - | 8 - 9 | 1699 | 138 | |
BX537872.1 | - | 8 - 9 | 1692 | 138 | |
KU178892.1 | - | 7 - 8 | 1471 | 138 | |
KU178894.1 | - | 6 - 7 | 1305 | 138 |