The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160147.1 | NP_001153619.1 | 6 - 7 | 1642 | 111 |
NM_001160148.1 | NP_001153620.1 | 5 - 6 | 1621 | 111 | |
NM_030637.2 | NP_085140.2 | 5 - 6 | 1621 | 111 | |
XM_005268102.2 | XP_005268159.1 | 6 - 7 | 1712 | 111 | |
XM_005268103.2 | XP_005268160.1 | 6 - 7 | 1631 | 111 | |
XM_005268105.2 | XP_005268162.1 | 4 - 5 | 1481 | 111 | |
XM_011537188.2 | XP_011535490.1 | 7 - 8 | 1733 | 111 | |
XM_011537189.2 | XP_011535491.1 | 7 - 8 | 1733 | 111 | |
XM_017021668.1 | XP_016877157.1 | 6 - 7 | 1712 | 111 | |
XM_017021669.1 | XP_016877158.1 | 4 - 5 | 1481 | 111 | |
GenBank mRNA | AB051492.1 | - | 2 - 3 | 1982 | 111 |
AK058137.1 | - | 3 - 4 | 324 | 111 | |
AK125372.1 | - | 6 - 7 | 1524 | 111 | |
AK125556.1 | - | 5 - 6 | 886 | 111 | |
BC030703.2 | - | 5 - 6 | 1575 | 111 |