Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UTP20
Entrez Gene ID
27340
Gene Name
UTP20, small subunit processome component
Gene Aliases
1A6/DRIM, DRIM
Chromosome Location
Chr.12: 101280127 - 101386619 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014503.2 NP_055318.2 53 - 54 7108 70
GenBank mRNA AF072718.1 - 11 - 12 1307 70
AJ006778.1 - 53 - 54 7096 70
AK000333.1 - 8 - 9 891 70
AK297739.1 - 4 - 5 309 70

View Full Product Details