Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTP20
Entrez Gene ID
27340
Gene Name
UTP20, small subunit processome component
Gene Aliases
1A6/DRIM, DRIM
Chromosome Location
Chr.12: 101280127 - 101386619 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014503.2 NP_055318.2 55 - 56 7425 136
GenBank mRNA AF072718.1 - 13 - 14 1624 136
AJ006778.1 - 55 - 56 7413 136
AK000333.1 - 10 - 11 1208 136
AK297739.1 - 6 - 7 626 136

View Full Product Details