Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTP20
Entrez Gene ID
27340
Gene Name
UTP20, small subunit processome component
Gene Aliases
1A6/DRIM, DRIM
Chromosome Location
Chr.12: 101280127 - 101386619 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014503.2 NP_055318.2 5 - 6 675 196
GenBank mRNA AJ006778.1 - 5 - 6 663 196
AK307934.1 - 5 - 6 676 196

View Full Product Details