The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005100.3 | NP_005091.2 | 2 - 3 | 402 | 86 |
XM_017011517.1 | XP_016867006.1 | 2 - 3 | 552 | 86 | |
GenBank mRNA | AB210003.1 | - | 1 - 2 | 335 | 86 |
AK307578.1 | - | 1 - 2 | 316 | 86 | |
BC000188.1 | - | 1 - 2 | 196 | 86 | |
BC015830.1 | - | 1 - 2 | 186 | 86 | |
BC022814.1 | - | 1 - 2 | 196 | 86 | |
BC046095.1 | - | 1 - 2 | 324 | 86 | |
BC047728.1 | - | 2 - 3 | 402 | 86 | |
U81607.1 | - | 1 - 2 | 353 | 86 |