The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006510.4 | NP_006501.1 | 7 - 8 | 1305 | 146 |
GenBank mRNA | AF230393.1 | - | 7 - 8 | 1180 | 146 |
AF230394.1 | - | 7 - 8 | 1180 | 146 | |
AJ420519.1 | - | 7 - 8 | 1245 | 146 | |
AX776009.1 | - | 7 - 8 | 1304 | 146 | |
AX776011.1 | - | 7 - 8 | 1304 | 146 | |
BC013580.1 | - | 7 - 8 | 1228 | 146 | |
BC066924.1 | - | 7 - 8 | 1276 | 146 | |
J03407.1 | - | 7 - 8 | 1180 | 146 |