The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004706.3 | NP_004697.2 | 20 - 21 | 2023 | 69 |
NM_198977.1 | NP_945328.1 | 19 - 20 | 1924 | 69 | |
NM_199002.1 | NP_945353.1 | 20 - 21 | 2048 | 69 | |
XM_005259386.4 | XP_005259443.1 | 22 - 23 | 2555 | 69 | |
XM_005259390.4 | XP_005259447.1 | 20 - 21 | 2371 | 69 | |
XM_011527468.2 | XP_011525770.1 | 21 - 22 | 2383 | 69 | |
GenBank mRNA | AL832294.1 | - | 11 - 12 | 1187 | 69 |
BC005155.2 | - | 17 - 18 | 1761 | 69 | |
BC011726.2 | - | 19 - 20 | 1899 | 69 | |
BC034013.1 | - | 20 - 21 | 1979 | 69 | |
BC035127.1 | - | 7 - 8 | 1612 | 69 | |
BC067262.1 | - | 19 - 20 | 1867 | 69 | |
BT007421.1 | - | 17 - 18 | 1737 | 69 | |
U64105.1 | - | 20 - 21 | 1968 | 69 |