The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004706.3 | NP_004697.2 | 21 - 22 | 2104 | 119 |
NM_198977.1 | NP_945328.1 | 20 - 21 | 2005 | 119 | |
NM_199002.1 | NP_945353.1 | 21 - 22 | 2129 | 119 | |
XM_005259386.4 | XP_005259443.1 | 23 - 24 | 2636 | 119 | |
XM_005259390.4 | XP_005259447.1 | 21 - 22 | 2452 | 119 | |
XM_011527468.2 | XP_011525770.1 | 22 - 23 | 2464 | 119 | |
GenBank mRNA | AL832294.1 | - | 12 - 13 | 1268 | 119 |
BC005155.2 | - | 18 - 19 | 1842 | 119 | |
BC011726.2 | - | 20 - 21 | 1980 | 119 | |
BC034013.1 | - | 21 - 22 | 2060 | 119 | |
BC035127.1 | - | 8 - 9 | 1693 | 119 | |
BC067262.1 | - | 20 - 21 | 1948 | 119 | |
BT007421.1 | - | 18 - 19 | 1818 | 119 | |
U64105.1 | - | 21 - 22 | 2049 | 119 |