Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA4
Entrez Gene ID
5267
Gene Name
serpin family A member 4
Gene Aliases
KAL, KLST, KST, PI-4, PI4, kallistatin
Chromosome Location
Chr.14: 94561420 - 94569913 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289032.1 NP_001275961.1 3 - 4 1329 75
NM_001289033.1 NP_001275962.1 3 - 4 1174 75
NM_006215.3 NP_006206.2 3 - 4 1014 75
GenBank mRNA AK309586.1 - 3 - 4 1307 75
AK313196.1 - 3 - 4 992 75
BC014992.1 - 3 - 4 988 75
BX248009.1 - 3 - 4 1089 75
BX248760.1 - 3 - 4 977 75
L19684.1 - 2 - 3 922 75

View Full Product Details