Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA4
Entrez Gene ID
5267
Gene Name
serpin family A member 4
Gene Aliases
KAL, KLST, KST, PI-4, PI4, kallistatin
Chromosome Location
Chr.14: 94561420 - 94569913 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289032.1 NP_001275961.1 4 - 5 1495 71
NM_001289033.1 NP_001275962.1 4 - 5 1340 71
NM_006215.3 NP_006206.2 4 - 5 1180 71
GenBank mRNA AK309586.1 - 4 - 5 1473 71
AK313196.1 - 4 - 5 1158 71
BC014992.1 - 4 - 5 1154 71
BX248009.1 - 4 - 5 1255 71
BX248760.1 - 4 - 5 1143 71
L19684.1 - 3 - 4 1088 71

View Full Product Details