The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000724.3 | NP_000715.2 | 11 - 12 | 1751 | 74 |
NM_001167945.1 | NP_001161417.1 | 12 - 13 | 1470 | 74 | |
NM_201570.2 | NP_963864.1 | 11 - 12 | 1605 | 74 | |
NM_201571.3 | NP_963865.2 | 12 - 13 | 1584 | 74 | |
NM_201572.3 | NP_963866.2 | 12 - 13 | 1512 | 74 | |
NM_201590.2 | NP_963884.2 | 11 - 12 | 1381 | 74 | |
NM_201593.2 | NP_963887.2 | 12 - 13 | 1247 | 74 | |
NM_201596.2 | NP_963890.2 | 12 - 13 | 1361 | 74 | |
NM_201597.2 | NP_963891.1 | 12 - 13 | 1289 | 74 | |
XM_005252588.3 | XP_005252645.1 | 11 - 12 | 1495 | 74 | |
XM_005252591.3 | XP_005252648.1 | 6 - 7 | 556 | 74 | |
XM_006717502.3 | XP_006717565.1 | 11 - 12 | 1414 | 74 | |
XM_011519659.2 | XP_011517961.1 | 11 - 12 | 1309 | 74 | |
XM_011519660.2 | XP_011517962.1 | 11 - 12 | 1852 | 74 | |
XM_017016625.1 | XP_016872114.1 | 12 - 13 | 1469 | 74 | |
GenBank mRNA | AB208917.1 | - | 11 - 12 | 1637 | 74 |
AF137376.1 | - | 12 - 13 | 1299 | 74 | |
AF137377.1 | - | 11 - 12 | 1142 | 74 | |
AF285239.2 | - | 11 - 12 | 1139 | 74 | |
AF423189.1 | - | 11 - 12 | 1136 | 74 | |
AF423190.1 | - | 12 - 13 | 1217 | 74 | |
AF423191.1 | - | 12 - 13 | 1301 | 74 | |
AF423192.1 | - | 11 - 12 | 1157 | 74 | |
AF465485.1 | - | 11 - 12 | 1031 | 74 | |
AK293937.1 | - | 11 - 12 | 1052 | 74 | |
AK296471.1 | - | 12 - 13 | 1268 | 74 | |
AK299873.1 | - | 11 - 12 | 1507 | 74 | |
AK307526.1 | - | 12 - 13 | 1470 | 74 | |
AY393858.1 | - | 12 - 13 | 1217 | 74 | |
AY393859.1 | - | 11 - 12 | 1139 | 74 | |
AY393860.1 | - | 12 - 13 | 1229 | 74 | |
AY393861.1 | - | 12 - 13 | 1145 | 74 | |
AY393862.1 | - | 11 - 12 | 1067 | 74 | |
AY675091.1 | - | 5 - 6 | 551 | 74 | |
BC136409.1 | - | 12 - 13 | 1324 | 74 | |
S60415.1 | - | 12 - 13 | 1361 | 74 | |
U95019.1 | - | 11 - 12 | 1637 | 74 |