The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006351.3 | NP_006342.2 | 4 - 5 | 566 | 84 |
GenBank mRNA | AF026030.1 | - | 4 - 5 | 383 | 84 |
AF041254.1 | - | 4 - 5 | 424 | 84 | |
AK223062.1 | - | 4 - 5 | 409 | 84 | |
AK289634.1 | - | 4 - 5 | 566 | 84 | |
AK309415.1 | - | 5 - 6 | 517 | 84 | |
BC033628.1 | - | 4 - 5 | 396 | 84 | |
X58234.1 | - | - | 104 | 84 |