The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024654.4 | NP_078930.3 | 11 - 12 | 2053 | 111 |
XM_005263493.3 | XP_005263550.1 | 9 - 10 | 1385 | 111 | |
XM_011542147.2 | XP_011540449.1 | 6 - 7 | 979 | 111 | |
XM_017002338.1 | XP_016857827.1 | 9 - 10 | 1563 | 111 | |
XM_017002339.1 | XP_016857828.1 | 8 - 9 | 1279 | 111 | |
GenBank mRNA | AK026976.1 | - | 7 - 8 | 1063 | 111 |
AK091284.1 | - | 11 - 12 | 1990 | 109 | |
AK289653.1 | - | 11 - 12 | 1985 | 111 | |
BC009257.2 | - | 8 - 9 | 1143 | 111 | |
BC012439.1 | - | 4 - 5 | 629 | 111 | |
BC063639.1 | - | 8 - 9 | 1093 | 111 | |
BC073860.1 | - | 7 - 8 | 1056 | 111 | |
BC094836.1 | - | 10 - 11 | 1428 | 111 | |
BC105095.1 | - | 11 - 12 | 2053 | 111 | |
BC110849.1 | - | 11 - 12 | 1962 | 111 | |
BC112278.1 | - | 11 - 12 | 2053 | 111 | |
BC143646.1 | - | 11 - 12 | 2085 | 111 | |
BX393201.2 | - | 1 - 2 | 111 |