The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256849.1 | NP_001243778.1 | 21 - 22 | 2796 | 114 |
NM_001308632.1 | NP_001295561.1 | 20 - 21 | 2802 | 114 | |
NM_002691.3 | NP_002682.2 | 21 - 22 | 2793 | 114 | |
XM_005259008.4 | XP_005259065.1 | 21 - 22 | 2753 | 114 | |
XM_011527038.1 | XP_011525340.1 | 21 - 22 | 2851 | 114 | |
XM_017026881.1 | XP_016882370.1 | 21 - 22 | 2870 | 114 | |
XM_017026882.1 | XP_016882371.1 | 21 - 22 | 2714 | 114 | |
GenBank mRNA | BC008800.1 | - | 21 - 22 | 2768 | 114 |
DQ234348.1 | - | 20 - 21 | 2802 | 114 | |
M80397.1 | - | 21 - 22 | 2777 | 114 | |
M81735.1 | - | 21 - 22 | 2766 | 114 |