Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLD1
Entrez Gene ID
5424
Gene Name
DAN polymerase delta 1, catalytic subunit
Gene Aliases
CDC2, CRCS10, MDPL, POLD
Chromosome Location
Chr.19: 50384291 - 50418018 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256849.1 NP_001243778.1 8 - 9 1045 87
NM_001308632.1 NP_001295561.1 7 - 8 973 87
NM_002691.3 NP_002682.2 8 - 9 1042 87
NR_046402.1 8 - 9 1042 87
XM_005259008.4 XP_005259065.1 8 - 9 1074 87
XM_011527038.1 XP_011525340.1 8 - 9 1100 87
XM_017026881.1 XP_016882370.1 8 - 9 1119 87
XM_017026882.1 XP_016882371.1 8 - 9 1035 87
GenBank mRNA AB209560.1 - 8 - 9 1161 87
BC008800.1 - 8 - 9 1017 87
DQ234348.1 - 7 - 8 973 87
M80397.1 - 8 - 9 1026 87
M81735.1 - 8 - 9 1015 87

View Full Product Details