Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK6
Entrez Gene ID
5653
Gene Name
kallikrein related peptidase 6
Gene Aliases
Bssp, Klk7, PRSS18, PRSS9, SP59, hK6
Chromosome Location
Chr.19: 50958631 - 50969673 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319948.1 NP_001306877.1 1 - 2 185 113
NM_002774.3 NP_002765.1 1 - 2 185 113
GenBank mRNA AF013988.1 - 1 - 2 87 113
AK314897.1 - 1 - 2 90 113
AY279383.1 - 1 - 2 186 113
D78203.1 - 1 - 2 96 113
U62801.1 - 1 - 2 186 113

View Full Product Details