Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK6
Entrez Gene ID
5653
Gene Name
kallikrein related peptidase 6
Gene Aliases
Bssp, Klk7, PRSS18, PRSS9, SP59, hK6
Chromosome Location
Chr.19: 50958631 - 50969673 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012964.2 NP_001012982.1 3 - 4 641 137
NM_001012965.2 NP_001012983.1 2 - 3 593 137
NM_002774.3 NP_002765.1 4 - 5 444 137
GenBank mRNA AF013988.1 - 4 - 5 346 137
AK314897.1 - 4 - 5 349 137
AY318867.1 - 3 - 4 450 137
AY318869.1 - 3 - 4 436 137
AY318870.1 - 2 - 3 388 137
BC015525.2 - 3 - 4 417 137
BT006852.1 - 2 - 3 200 137
D78203.1 - 4 - 5 355 137
DQ223012.1 - 2 - 3 292 137
U62801.1 - 4 - 5 445 137

View Full Product Details