Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK11
Entrez Gene ID
11012
Gene Name
kallikrein related peptidase 11
Gene Aliases
PRSS20, TLSP
Chromosome Location
Chr.19: 51022231 - 51028034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136032.2 NP_001129504.1 4 - 5 572 77
NM_001167605.1 NP_001161077.1 4 - 5 659 77
NM_006853.2 NP_006844.1 4 - 5 584 77
NM_144947.1 NP_659196.1 4 - 5 666 77
XM_011526369.1 XP_011524671.1 4 - 5 810 77
XM_011526370.2 XP_011524672.1 4 - 5 684 77
XM_011526371.2 XP_011524673.1 5 - 6 804 77
XM_011526372.2 XP_011524674.1 4 - 5 717 77
XM_011526373.1 XP_011524675.1 4 - 5 587 77
GenBank mRNA AB012917.1 - 4 - 5 579 77
AB013730.1 - 4 - 5 578 77
AB041036.1 - 4 - 5 666 77
AB078780.1 - 3 - 4 579 77
AB259014.1 - 4 - 5 570 77
AB261897.1 - 3 - 4 579 77
AY359014.1 - 4 - 5 563 77
BC022068.1 - 4 - 5 590 77
BG697071.1 - 4 - 5 572 77

View Full Product Details