Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KHSRP
Entrez Gene ID
8570
Gene Name
KH-type splicing regulatory protein
Gene Aliases
FBP2, FUBP2, KSRP
Chromosome Location
Chr.19: 6413104 - 6424811 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003685.2 NP_003676.2 19 - 20 2238 149
XM_017027409.1 XP_016882898.1 20 - 21 2017 149
GenBank mRNA U69126.1 - 19 - 20 1952 146
U94832.1 - 19 - 20 2221 149

View Full Product Details