The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003685.2 | NP_003676.2 | 3 - 4 | 492 | 105 |
XM_005259668.4 | XP_005259725.1 | 3 - 4 | 382 | 105 | |
XM_011528395.2 | XP_011526697.1 | 4 - 5 | 347 | 105 | |
XM_017027408.1 | XP_016882897.1 | 4 - 5 | 346 | 105 | |
XM_017027409.1 | XP_016882898.1 | 4 - 5 | 346 | 105 | |
XM_017027410.1 | XP_016882899.1 | 3 - 4 | 241 | 105 | |
GenBank mRNA | AK300957.1 | - | 4 - 5 | 360 | 105 |
BC085004.1 | - | 3 - 4 | 456 | 105 | |
DC388502.1 | - | 4 - 5 | 360 | 105 | |
U69126.1 | - | 3 - 4 | 215 | 105 | |
U94832.1 | - | 3 - 4 | 475 | 105 |