Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C3
Entrez Gene ID
718
Gene Name
complement component 3
Gene Aliases
AHUS5, ARMD9, ASP, C3a, C3b, CPAMD1, HEL-S-62p
Chromosome Location
Chr.19: 6677835 - 6720682 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000064.3 NP_000055.2 12 - 13 1570 65
GenBank mRNA AK304071.1 - 12 - 13 1538 65
BC150179.1 - 12 - 13 1509 65
BC150200.1 - 12 - 13 1509 65
EU794602.1 - 12 - 13 1509 65
K02765.1 - 12 - 13 1537 65

View Full Product Details