The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000064.3 | NP_000055.2 | 34 - 35 | 4355 | 113 |
GenBank mRNA | AK094728.1 | - | 1 - 2 | 1222 | 113 |
BC150179.1 | - | 34 - 35 | 4294 | 113 | |
BC150200.1 | - | 34 - 35 | 4294 | 113 | |
EU794602.1 | - | 34 - 35 | 4294 | 113 | |
K02765.1 | - | 34 - 35 | 4322 | 113 |