The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000635.3 | NP_000626.2 | 9 - 10 | 1290 | 59 |
NM_134433.2 | NP_602309.1 | 8 - 9 | 1215 | 59 | |
XM_011528171.2 | XP_011526473.1 | 10 - 11 | 1256 | 59 | |
XM_011528172.1 | XP_011526474.1 | 9 - 10 | 1113 | 59 | |
XM_017027107.1 | XP_016882596.1 | 8 - 9 | 1140 | 59 | |
XM_017027108.1 | XP_016882597.1 | 7 - 8 | 1065 | 59 | |
GenBank mRNA | AK024288.1 | - | 6 - 7 | 829 | 59 |
AK291196.1 | - | 8 - 9 | 1094 | 59 | |
AK302661.1 | - | 3 - 4 | 278 | 59 | |
AK303321.1 | - | 6 - 7 | 599 | 59 | |
BC028579.1 | - | 9 - 10 | 1202 | 59 | |
BC071571.1 | - | 9 - 10 | 1171 | 59 | |
BX648130.1 | - | 9 - 10 | 1125 | 59 | |
X76091.1 | - | 9 - 10 | 1180 | 59 |