Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALKBH7
Entrez Gene ID
84266
Gene Name
alkB homolog 7
Gene Aliases
ABH7, SPATA11, UNQ6002
Chromosome Location
Chr.19: 6372433 - 6375250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032306.3 NP_115682.1 3 - 4 891 91
XM_005259658.4 XP_005259715.1 3 - 4 467 91
GenBank mRNA AK223336.1 - 3 - 4 514 91
AK311956.1 - 3 - 4 530 91
AY358858.1 - 3 - 4 891 91
AY427650.2 - 3 - 4 891 91
BC004393.1 - 3 - 4 546 91
BM271861.1 - 1 - 2 91

View Full Product Details