Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASXL2
Entrez Gene ID
55252
Gene Name
additional sex combs like 2, transcriptional regulator
Gene Aliases
ASXH2
Chromosome Location
Chr.2: 25733758 - 25878485 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018263.4 NP_060733.4 10 - 11 1258 66
XM_006712039.3 XP_006712102.1 6 - 7 3379 66
XM_006712040.2 XP_006712103.1 6 - 7 665 66
XM_011532950.2 XP_011531252.1 9 - 10 1297 66
XM_011532951.2 XP_011531253.1 8 - 9 985 66
XM_017004429.1 XP_016859918.1 10 - 11 1088 66
XM_017004430.1 XP_016859919.1 7 - 8 3445 66
GenBank mRNA AB051472.2 - 10 - 11 1258 66
AB084279.1 - 10 - 11 1121 66
BC042999.2 - 6 - 7 696 66
BX647527.1 - 11 - 12 1331 66
CR933613.1 - 10 - 11 1302 66

View Full Product Details