Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTSW
Entrez Gene ID
1521
Gene Name
cathepsin W
Gene Aliases
LYPN
Chromosome Location
Chr.11: 65879813 - 65883741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335.3 NP_001326.2 7 - 8 789 85
GenBank mRNA AF013611.1 - 7 - 8 747 85
BC035637.1 - 7 - 8 752 85
BC048255.1 - 7 - 8 789 85

View Full Product Details